missing translation for 'onlineSavingsMsg'
Learn More

JULABO™ Basket for 1001F

JULABO Basket for 1001F is for use with refrigerated circulators CORIO CD-1001F, CORIO CP-1001F, or DYNEO DD-1001F.

Brand:  JULABO™ 9970337

Product Code. 17128538

  • 5510.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stainless steel basket holds 20 bottles, 0.33 to 0.5 L
Specifications

Specifications

20 Bottles, 0.33 L to 0.5 L each
Basket
1 Pk.
1001 F
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers