Learn More
BD BBL™ Dehydrated Culture Media: MacConkey II Agar
For isolation of gram-negative enteric bacilli from specimens that may contain swarming strains of Proteus
Brand: BD 212306
This item is not returnable.
View return policy
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, MCM9, SMD, SMWW, USP
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO 9001:2008, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, MCM9, SMD, SMWW, USP | |
Dehydrated | |
Agar | |
Isolating of gram negative enteric Bacilli from specimens that may contain swaming strains of proteus | |
500 g |
Refrigerate | |
MacConkey II Agar | |
Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L | |
Bottle |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1A
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : oxgall
Product Content Correction
Product Title
BD BBL™ Dehydrated Culture Media: MacConkey II Agar > MacConkey II Agar; 500g
Spot an opportunity for improvement?Share a Content Correction