Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II Agar w/MUG Prepared Plated Media
Two media formulations in single plate for cultivating fastidious microorganisms
Brand:  BD 221949
Product Code. 17372351
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
TSA II
  • For the visualization of hemolytic reactions produced by many bacterial species
MacConkey II Agar with MUG
  • For presumptive identification of Escherichia coli
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO/DIS, 11014
Liquid
Trypticase Soy Agar with 5% Sheep Blood (TSA II)/MacConkey II Agar with MUG
Cultivation of fastidious microorganisms and visualization of hemolytic reactions
20/Pk.
Dark Red
United States
Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.1g/L, Agar 13.5g/L
Plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II Agar w/MUG Prepared Plated Media > Trypticase Soy Agar with 5% Sheep Blood (TSA II)/MacConkey II Agar with MUG; 20/Pk.