Learn More
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II Agar w/MUG Prepared Plated Media
Two media formulations in single plate for cultivating fastidious microorganisms
Brand: BD 221949
This item is not returnable.
View return policy
Description
TSA II
- For the visualization of hemolytic reactions produced by many bacterial species
- For presumptive identification of Escherichia coli
Specifications
ISO/DIS, 11014 | |
Liquid | |
Trypticase Soy Agar with 5% Sheep Blood (TSA II)/MacConkey II Agar with MUG | |
Cultivation of fastidious microorganisms and visualization of hemolytic reactions | |
20/Pk. |
Dark Red | |
United States | |
Pancreatic digest of Gelatin 17g/L, Pancreatic digest of Casein 1.5g/L, Peptic digest of Animal Tissue 1.5g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Neutral red 0.03g/L, Crystal Violet 0.001g/L, 4-methylumbelliferyl-beta-D-glucuronide 0.1g/L, Agar 13.5g/L | |
Plate |
Product Content Correction
Product Title
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II Agar w/MUG Prepared Plated Media > Trypticase Soy Agar with 5% Sheep Blood (TSA II)/MacConkey II Agar with MUG; 20/Pk.
Spot an opportunity for improvement?Share a Content Correction