Learn More
BD Difco™ Dehydrated Culture Media: EE Broth Mossel Enrichment
For selectively enriching and detecting Enterobacteriaceae, particularly from foods
Brand: BD 256620
Description
- Formulation: Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L, Oxgall 20g/mL
Certifications
CE, COMPF, EP, ISO, JP, USP
Specifications
CE, COMPF, EP, ISO, JP, USP | |
Dehydrated | |
Microbiology Media | |
For selectively enriching and detecting Enterobacteriaceae, particularly from food | |
500 g |
<30°C | |
EE Broth | |
Pancreatic digest of Gelatin 10g/L, Dextrose 5g/L, Disodium Phosphate 8g/L, Monopotassium Phosphate 2g/L, Brilliant green 15mg/L, Oxgall 20g/mL | |
Bottle |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P330-Rinse mouth.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : oxgall