missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Urea Broth
For detection of urease producing bacteria
Brand: BD 227210
This item is not returnable.
View return policy
Description

Specifications
AOAC, BAM, COMPF, SMD | |
Dehydrated | |
Microbiology Media | |
For differentiation of organisms, especially the Enterobacteriaceae, on the basis of Urease production | |
500 g |
Refrigerate | |
Urea Broth | |
Yeast extract 0.1g/L, Monopotassium Phosphate 9.1g/L, Dipotassium Phosphate 9.5g/L, Urea 20g/L, Phenol red 0.01g/L | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: Urea Broth > Urea Broth; 500g
Spot an opportunity for improvement?Share a Content Correction