missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
- Plates: 2 Pre-coated ELISPOT plates
- Biotinylated Detection Antibody
- Enzyme Conjugate (Streptavidin-HRP)
- Assay Diluent
- Wash Concentrate (20)
- PBS (10)
- AEC Substrate Buffer
- AEC Chromogen
- Certificate of Analysis, with lot-specific optimal dilution instructions
ELISPOT assay derives its specificity and sensitivity by employing high affinity capture and detection antibodies and enzyme-amplification. Although originally developed for analyzing specific antibody-secreting cells, 2,3 the assay has been adapted for measuring the frequencies of cells that produce and secrete other effector molecules, such as cytokines.
- Cells analyzed by ELISPOT can be transferred for cloning, proliferation assays, flow cytometry, or other methods of analysis

Specifications
Specifications
Antigen | IFN-gamma |
Cross Reactivity | Human |
Includes | Antibody coated plate, Biotinylated Human IFN-g detection Antibody, Streptavidin HRP, wash concentrate, PBS (10X) concentrate, assay diluent, AEC Substrate Buffer, AEC chromogen |
Quantity | 2 Plates |
Content And Storage | 4°C |
For Use With (Application) | Cloning, proliferation assays, flow cytometry or other methods of analysis |
Product Type | Human IFN-gamma ELISPOT kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Human IFN-Îł ELISPOT Kit >
Spot an opportunity for improvement?Share a Content Correction