missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Compatible with the entire BD Vacutainer system of blood collection products
- Standard size, clear
- Non-sterile
- Clear
- Single use, non-stackable
- Latex free
Compliance
Complies with OSHA Safety and Health Information Bulletin (SHIB) on the disposal of contaminated needles and blood tube holders used for phlebotomy

Specifications
Specifications
Color | Clear |
Disposable | Single-use |
For Use With (Application) | Reduces the hazards of contact with Blood borne pathogens compared to reusable holders |
Latex Free | Yes |
Size | Standard |
Product Type | Tube Holder |
Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Vacutainer™ Tube Holder >
Spot an opportunity for improvement?Share a Content Correction