Learn More
H-B Instrument™ DURAC™ Single Channel Three Button Timer with Memory
Single channel timer has three key operation for ease of use
Brand:  H-B Instrument™ B61700-3400
127.81 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 17693125
  • 139.00 DKK / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Includes: One AAA battery
Description
Description
  • Timing capacity: 99 minutes: 59 seconds
  • Count direction: up/down
  • Last time set memory
  • Audible alarm; displays elapsed time after countdown
  • Mounting options: lay flat on bench, stand on edge for vertical view, magnetic back for metal surface attachment
  • Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Specifications
Specifications
Single Channel Timer
±5 seconds/day
1 sec.
10/Cs.
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
H-B Instrument™ DURAC™ Single Channel Three Button Timer with Memory > Single channel three button timer with memory