Learn More
Bel-Art™ SP Scienceware™ Riteflow™ Flowmeter Leveling Base Kit
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
Brand:  Bel-Art™ H40404-1000
Product Code. 10743694
  • 1323.88 DKK / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Description
  • 15.2cm (6 in.) Plexiglas™ triangular base
  • Three adjustable feet
  • Vial level
  • For use with Riteflow mounted flowmeters
Specifications
Specifications
Scienceware Flowmeters Catalog Nos.: 11-163-75 and -77 Series
Leveling Base Kit
Assures that the flowmeter is exactly vertical eliminating error caused by friction between the float and the flow tube
Documents
One moment while we fetch your results.
Product Title
Bel-Art™ SP Scienceware™ Riteflow™ Flowmeter Leveling Base Kit > Leveling base kit