Learn More
Lenz Laborglasinstrumente™ Adapter with NS Stopcock, Bent
For distillation tubing connections. Lenz Laborglasinstrumente™ Adapter with NS Stopcock, Bent are manufactured from durable DURAN™ glass tubing.
308.00 DKK - 319.00 DKK
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10460513
|
Lenz Laborglasinstrumente™
05098002 |
NS 19/26 |
308.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10359860
|
Lenz Laborglasinstrumente™
05098003 |
NS 24/29 |
319.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Bent glass olive at 90° angle
- Bore: 2.5mm
- Stopcock with solid glass plug
- Temperature and chemical resistance
Specifications
Bent Adapter with NS Stopcock | |
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Adapter with NS Stopcock, Bent