Learn More
Benzhydrazide, 98%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 113°C to 117°C |
Quantity | 100 g |
Beilstein | 471797 |
Formula Weight | 136.15 |
Percent Purity | 98% |
Chemical Name or Material | Benzhydrazide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253015 | Certificate of Analysis | 26/11/2024 | A12268.14,A12268.36,A12268.22 |
10231895 | Certificate of Analysis | 20/12/2023 | A12268.14,A12268.36,A12268.22 |
10241439 | Certificate of Analysis | 27/06/2022 | A12268.14,A12268.36,A12268.22 |
10186901 | Certificate of Analysis | 22/11/2021 | A12268.14,A12268.36,A12268.22 |
10223968 | Certificate of Analysis | 22/11/2021 | A12268.14,A12268.36,A12268.22 |
Safety and Handling


- Benzhydrazide
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Benzoic acid, hydrazide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.