missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (BBM.1), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP2-81045AF405
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
beta 2-Microglobulin | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
B2M | |
Molt 4, a human T cell line. | |
0.1 mL | |
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
567 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
BBM.1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Binds human beta 2-Microglobulin, a component of MHC class I molecules, which are present on all nucleated cells (excludes red blood cells) and involved in the presentation of peptide antigens to the immune system. | |
Store at 4°C in the dark. | |
IgG κ |