missing translation for 'onlineSavingsMsg'
Learn More
beta-Propiolactone, 98%, pure, Thermo Scientific Chemicals Product Code.: 10625511

beta-Propiolactone, 98%, pure, Thermo Scientific Chemicals

Product Code. 10625511 Shop All Thermo Scientific Chemicals Products
25 mL, Glass bottle
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10625511

Brand: Thermo Scientific Acros 269040250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 57-57-8 | C3H4O2 | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Packaging Note: Glass bottle is put into a can for shipping.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-57-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00005169
InChI Key VEZXCJBBBCKRPI-UHFFFAOYSA-N
Synonym beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolaktonShow MoreShow Less
PubChem CID 2365
ChEBI CHEBI:49073
IUPAC Name oxetan-2-one
SMILES O=C1CCO1

Specifications

Density 1.1460g/mL
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 mL
Formula Weight 72.06
Grade Pure
Chemical Name or Material β-Propiolactone, Pure

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0438090 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number269041000,269040050,269040250,269045000
Lot NumberA0444258 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number269041000,269040050,269040250,269045000
Lot NumberA0441579 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog Number269041000,269040050,269040250,269045000
Lot NumberA0438090 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number269041000,269040050,269040250,269045000
Lot NumberA0437091 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number269041000,269040050,269040250,269045000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
beta-Propiolactone, 98%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.