Learn More
Fisherbrand™ Beta Shields
Cleaver Scientific radiation shielding for protection from Beta radiation.
Brand:  Fisherbrand™ CSR-S1OT
 View more versions of this product
Product Code. 12347177
  • 3710.00 DKK / Each
Only null left
Estimated Shipment: 08-12-2025
Add to basket
Description
Description
  • Available in two sizes, with either a flat base to provide bench
  • Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
  • A range of angles offers increased manueverability, while clear optical acrylic aids visualisation
  • Beta - Small Fixed 45° Angle - Curved Base 30 x 45cm
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
Specifications
Specifications
45° Angle
Beta Shield
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Beta Shields > Angle: 45°