Learn More
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots
Cleanroom Coverall made of CleanTough™ fabric for lightweight, low-linting body protection with front zip and protective flap to provide contaminant and liquid splash barrier.
Brand: Ansell S-BDFC-XS
85.58 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reduced contamination risks: Lightweight CleanTough™ material means the BioClean-D Coverall with Hood and Integrated Boots – Sterile S-BDFC is low-linting and ISO cleanroom-compliant, reducing contamination risks
- Optimized protection: This sterile coverall features a front zip with a protective flap, resisting liquid splashes and chemicals*, while thumb loops and an elasticated hood, back, cuffs and ankles ensure a secure fit
- Enhanced features: This antistatic coverall features integrated boots and slip-resistant soles, for added workplace safety in laboratory settings
- Sterility assurance level: Special packaging eliminates cross contamination, while gamma irradiation sterilization ensures a minimum SAL of 10-6
- Contact customer service for chemotherapy drug permeation times and recommendations.
Specifications
Cleanroom Coveralls | |
Unisex | |
White | |
Standard | |
Elastic | |
Attached, Elastic Face | |
Bound | |
Anti-static, Chemicals, Liquid, Particle | |
ASTM F739, CE 0598, EN 1149-5:2008, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 13688:2013, ISO 11137-1:2006, EN ISO 14325, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009 | |
None | |
Class 10 (ISO 4) |
X-Small | |
Flashspun Polyethylene | |
Single-use | |
Attached Boots, Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Sterile | |
Hooded | |
No | |
Blending, compounding solids or liquids, Crime Scene Investigation (CSI), Forensics, Handling of Powders, Aseptic Filling and Labelling, Cleanroom Cleaning and Preparing, Medical Device Manufacturing, Biotechnologies Blending Operations, Handling Active Material, Maintaining Equipment and Instruments |
Product Content Correction
Product Title
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots > Sterility: Sterile
Spot an opportunity for improvement?Share a Content Correction