Learn More
Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Color:White, Sterility:Sterile; Lid:With Lid
Brand:  Corning™ 356693
Product Code. 10192822
  • 11338.00 DKK / Pack of 80
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Superior lot-to-lot and intra-well consistency
  • Minimized cross talk
  • Stackable design
  • Color: White
  • Bottom: Clear, flat, with lid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
20/Pk., 80/Cs.
BioCoat
96
Poly-D-Lysine
80
8 x 12
White
Polystyrene
Sterile
Flat Bottom (F-well)
80/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate > 96 wells, PDL treated, White, Sterile