missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Biohazard Bags, Irradiated

Made from FDA-compliant LDPE, each Biohazard Bag displays a large biohazard symbol and warning message. Supplied irradiated [minimum dose: 25 kGy (SAL: 10-6)] they are ideal for aseptic suites in cell therapy and related disciplines. Available flat packed in groups of 5 or on a plastic core roll.

3075.00 DKK - 4229.44 DKK

Specifications

Certifications/Compliance FDA: Regulation 21 CFR, 177.1520, 3.1; DOT: ASTM D1709 and ASTM D1922; Manufacturing: ISO 9001; Controlled Environments: ISO 5 - 9 Cleanrooms
Closure Type None
Design Biohazard Symbol and Warning Printed on Each Bag
Disposable Single-use
Quantity 50/Cs.
View More Specs

Products 2
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
17843818
View Documents
Micronova™
BL4-32YEIRBB
Yellow 120 L
3075.00 DKK
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
18886070
View Documents
Micronova™
BL4-4060REIRBB
Red 200 L
4229.44 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Product Type: Biohazard Bag
  • Material: LDPE
  • Thickness: 4 mm
  • Sterile: Yes
  • Sterilization Method: Irradiation [Minimum Dose: 25 kGy (SAL: 10-6)]
Specifications

Specifications

FDA: Regulation 21 CFR, 177.1520, 3.1; DOT: ASTM D1709 and ASTM D1922; Manufacturing: ISO 9001; Controlled Environments: ISO 5 - 9 Cleanrooms
Biohazard Symbol and Warning Printed on Each Bag
50/Cs.
Not Autoclavable
4 mm
None
Single-use
Sterile
Biohazardous Waste Disposal
5 Liners/Double Bag
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ Biohazard Bags, Irradiated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.