Learn More
Sartorius™ Biohit™ Proline Single-Channel Mechanical Pipettes
Ideal for universities and colleges or any laboratory seeking cost-efficient, reliable liquid-handling tool. Sartorius™ Biohit™ Proline Single-Channel Mechanical Pipettes are adjustable-range pipettes that can be used with many tip brands.
Brand:  Sartorius™ 726405
 View more versions of this product
Product Code. 11639152
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Light weight with high accuracy and precision for use by professionals
  • Wide range of pipettes with adjustable or fixed volume ranges
  • Easy volume setting with click stop mechanism
  • Relaxed grip with ergonomic finger support
  • Reduced risk of contamination with Safe-Cone Filters available
  • Compatible with many universal tips, including Optifit Tips and SafetySpace Tips
Proline Plus models feature:
  • Color coding on the pipette
  • Fully autoclavable, with only 3 parts to clear
  • More ergonomic design for lower required pipetting force
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO
Autoclavable
1
Sterile
1000 μL
1 Each
82 g
Optifit
Proline Plus
Auto
Mechanical Pipette
Ergonomic
Chemical