Learn More
Sartorius™ Biohit™ Tip Cone Housing
Sartorius™ Biohit™ Tip Cone Housing are designed for use with a variety of Proline manual and electronic multichannel pipettes. Available in several sizes for a variety of pipette models/tips.
Brand: Sartorius™ 726133
This item is not returnable.
View return policy
Description
- Made of durable thermoplastic
- Houses disposable tips for multi-channel pipettes
- Available in several sizes for a variety of mechanical and electronic units
Specifications
Thermoplastic | |
Tip Cone Housing | |
8/300μL Mline Multi Channel Pipette |