Learn More
Sartorius™ Biohit™ Tip Ejector Collars
Allows safe and easy pipette tip removal without touching the tips by hand. Sartorius™ Biohit™ Tip Ejector Collars are designed for use with a variety of mechanical and electronic single- or multi-channel pipette models/sizes.
Brand:  Sartorius™ 726051
 View more versions of this product
Product Code. 11604732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Removes pipette tips without touching the tips by hand
  • Activates a spring loaded tip cone
  • Tips are ejected with a simple thumb press on the ejector button
  • Compatible with Biohit&trade tips, including Optifit tips and SafetySpace™ Filter tips
  • Available in several sizes for a variety of mechanical and electronic units
  • Made of durable thermoplastic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tip Ejector Collar
3μL, 10μL mLINE Single Channel Pipette, Also fits 10μL Sealpette Pro Pipette