Learn More
Sartorius™ Biohit™ Tip Ejector Collars
Allows safe and easy pipette tip removal without touching the tips by hand. Sartorius™ Biohit™ Tip Ejector Collars are designed for use with a variety of mechanical and electronic single- or multi-channel pipette models/sizes.
Brand: Sartorius™ 721055
This item is not returnable.
View return policy
Description
- Removes pipette tips without touching the tips by hand
- Activates a spring loaded tip cone
- Tips are ejected with a simple thumb press on the ejector button
- Compatible with Biohit&trade tips, including Optifit tips and SafetySpace™ Filter tips
- Available in several sizes for a variety of mechanical and electronic units
- Made of durable thermoplastic
Specifications
Metal Blue | |
Tip Ejector Collar | |
1000μL Proline Mechanical Pipette |