Learn More
Sartorius™ Biosart™ Monitor
Microbiological and analytical filtration units offering superior performance
Brand: Sartorius™ 16403-47-04----K
1425.03 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
Specifications
Gray/White | |
48Pack | |
2 layers of 24 | |
Monitor |
Sterile | |
48Pack | |
48Pack |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%