Learn More
Bismuth rod, 12.7mm (0.5in) dia, 99.999+% (metals basis), Thermo Scientific Chemicals Product Code.: 11358747
Bismuth rod, 12.7mm (0.5in) dia, 99.999+% (metals basis), Thermo Scientific Chemicals
Product Code. 11358747
25 g
Click to view available options
Quantity:
25 g
100 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11358747
Brand: Thermo Scientific Alfa Aesar 035854.14
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]
Specifications
Quantity 25 g
Percent Purity ≥99.999%
Odor Odorless
Weight ≈12.5g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Bismuth rod, 12.7mm (0.5 in.) dia.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ20W006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035854.14,035854.22
Lot NumberY20G043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035854.14,035854.22
Lot NumberX11C039 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number035854.14,035854.22
Lot NumberW27C033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035854.14,035854.22
Lot NumberJ22U019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number035854.14,035854.22
RUO – Research Use Only
Product Content Correction
Product Title
Bismuth rod, 12.7mm (0.5in) dia, 99.999+% (metals basis), Thermo Scientific Chemicals >