Learn More
Brand™ BLAUBRAND™ 250mL Measuring Flask
With single certificate.
Brand: Brand™ 937251
383.12 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from Boro 3.3 glass with a polypropylene stopper
- Marks and inscriptions in high contrast blue enamel
- Wide neck
Specifications
DIN EN ISO 1042 | |
250 mL | |
Blue Enamel | |
Stopper |
Polypropylene | |
Borosilicate Glass (3.3) | |
Yes |
Product Content Correction
Product Title
Brand™ BLAUBRAND™ 250mL Measuring Flask >
Spot an opportunity for improvement?Share a Content Correction