Learn More
BRAND™ BLAUBRAND™ Bang Pattern Micro Burette
Shop All Brand Products
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 2
Description
Specifications
Specifications
Capacity (Metric) | 5 mL |
Material | Borosilicate Glass (3.3) |
Graduations | 0.01 mL |
No. per Pack | 2 |
Tolerance | 0.01 mL |
Class | Class AS |
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Bang Pattern Micro Burette >
Spot an opportunity for improvement?Share a Content Correction