Learn More
BRAND™ Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders
Constructed of borosilicate glass and complies with DIN EN ISO 4788 standards. BRAND™ Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders include integral spout and hexagonal glass base for stability. DE-M Marking amber stain graduations provide quick and easy volume assessment.
186.00 DKK - 979.00 DKK
Specifications
Base Type | Hexagonal |
---|---|
Calibration | To Contain |
Certifications/Compliance | DIN EN ISO 4788 |
Class | A |
Type | Graduated Measuring Cylinder |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | |||||
11824000
|
BRAND™
32754 |
500 mL | 5 mL |
744.00 DKK
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||||
11893990
|
BRAND™
32720 |
25 mL | 0.5 mL |
195.00 DKK
Pack of 2 |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
11804000
|
BRAND™
32728 |
50 mL | 1 mL |
215.00 DKK
Pack of 2 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
11883990
|
BRAND™
32708 |
10 mL | 0.2 mL |
186.00 DKK
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
10339710
|
BRAND™
32764 |
2000 mL | 20 mL |
979.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
10024070
|
BRAND™
32705 |
5 mL | 0.1 mL |
186.00 DKK
Pack of 2 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
|||||
10742321
|
BRAND™
32762 |
1000 mL | 10 mL |
580.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||||
10657851
|
BRAND™
32738 |
100 mL | 1 mL |
267.00 DKK
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||||
11814000
|
BRAND™
32748 |
250 mL | 2 mL |
436.00 DKK
Pack of 2 |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Class A tall form measuing cylinder constructed of borosilicate glass
- Complies with DIN EN ISO 4788 standards
- Autoclavable
- Calibrated to contain (TC, In).
- With integral spout and hexagonal glass base for stability
- DE-M Marking amber stain graduations provide quick and easy volume assessment
- USP individual certificate or DAkkS calibration certificate available upon request
Specifications
Hexagonal | |
DIN EN ISO 4788 | |
Graduated Measuring Cylinder | |
Round | |
Batch Certificate |
To Contain | |
A | |
Amber | |
Borosilicate Glass | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Blaubrand™ ETERNA Class A Borosilicate Glass Tall Form Measuring Cylinders