missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Batch Certificate

Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. Calibrated to contain a precise amount of liquid at a specific temperature. Includes polypropylene stopper and DE-M marking. Total order delivered from one batch (to the extent possible).

285.00 DKK - 400.00 DKK

Specifications

Calibration Calibrated To Contain
Closure Material Polypropylene
Color Clear
Material Borosilicate Glass
No. per Case 2 Pack
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15831508
View Documents
BRAND™
236838
5 mL
285.00 DKK
Pack of 2
Estimated Shipment: 02-06-2025
to see stock.
Add to basket
 
15841508
View Documents
BRAND™
236850
200 mL
400.00 DKK
Pack of 2
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
Description

Description

  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • Includes polypropylene stopper and DE-M marking
  • Total order delivered from one batch (to the extent possible)
Specifications

Specifications

Calibrated To Contain
Clear
2 Pack
A
Polypropylene
Borosilicate Glass
Yes
Stopper
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Blaubrand™ ETERNA™ Class A Volumetric Flasks, with Batch Certificate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.