Learn More
mb2 Chair on Castors - Blue Fabric Upholstered - Mid Lift Product Code.: 11987834
Estimated Shipment: 13-08-2025
Only null left
Add to basket
mb2 Chair on Castors - Blue Fabric Upholstered - Mid Lift
Product Code. 11987834
Middle lift
Click to view available options
:
Middle lift
Product Code. 11987834
Brand: mb2 AT1B2D1NE2D6016
Only null left
Estimated Shipment: 13-08-2025
Add to basket
Only null left
Add to basket
Chair on castors - Mid lift - Permanent contact mechanism - blue fabric upholstered
  • Chair in fabric black
  • Seat and back in blue fabric upholstered
  • Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
  • Pneumatic cylinder mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors.
Specifications
Product Type Contact Seat Chair
Color Blue
Adjustable (Height) Yes
Adjustable Height
Back Style Ergonomic
Height (Metric) 700 mm
Holds 5 Wheels
Quantity 1 Ea.
Seating Height 490 to 690 mm
Weight (Metric) 9.86 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt Yes
Material Fabric
Seating Type Medium Bench Height
Show More Show Less
Product Content Correction
Product Title
mb2 Chair on Castors - Blue Fabric Upholstered - Mid Lift >