Learn More
Scotch-Brite™ Blue Floor Pads
Designed for frequent burnishing with less dust and burning, used to restore the gloss to dull floors. Can be used wet for regular maintenance of unprotected floors. These pads are also recommended for dry stripping soft floors.
Brand:  Scotch-Brite™ 7100182687
 View more versions of this product
Product Code. 17128830
  • 514.00 DKK / Pack of 5
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Description
  • Heavy-duty scrub pad removes dirt, spills, scuffs, and 1-2 coats of finish
  • Leaves surfaces clean for recoating
  • Designed for frequent burnishing with less dust and burning
  • Washable and reusable pad is economical and helps postpone end-of-life disposal to landfills
  • Versatile pads can be used for heavy-duty spray cleaning and foam scrubbing
  • Available in sizes for rotary and automatic floor machines running at low speed (175-600 rpm)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stripping Pads
280 mm
Rotary and Automatic Floor Machines
Blue
Spray Cleaning, and Foam Scrubbing
Round
Product Title
Scotch-Brite™ Blue Floor Pads > 280 mm