Learn More
ABEBA™ Blue Autoclavable Clogs 9610
Antistatic and autoclavable at 121°C. High slip resistance.
Brand: ABEBA™ 9610-37/38
This item is not returnable.
View return policy
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- Blue
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
CE EN ISO 20347:2012 OB A, E SRC | |
9610 | |
Controlled Environments Shoes |
Blue | |
Static |