Learn More
Fisherbrand™ Blue Micro PTFE Stir Bar
Fisherbrand™ Blue Micro PTFE Stir Bar is an inert and leak-free stirrer for a wide variety of applications where identification is important.
Brand:  Fisherbrand™ 001.806-B
Product Code. 16245841
  • 56.35 DKK / Pack of 10
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Micro stirrer bars
  • For use where identification is of prime importance
  • PTFE coatings are not as inert as pure PTFE
  • Individually polished to stay cleaner for longer
Specifications
Specifications
Autoclavable
Blue
Containers & Vessels / Stirrer Hot Plates
6 mm
Micro
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
3 mm
10/Pk.