Learn More
Sampling Systems™ Blue SteriWare™ Scrapers
The risk of cross contamination is eliminated by using Blue SteriWare™ Scrapers. Cleanroom manufactured and available sterile, these funnels are ideal for scraping and cleaning in the food, cosmetic, and pharmaceutical industry. Blue color allows you to see the scraper if left in product.
1345.00 DKK - 1595.00 DKK
Specifications
Dimensions (L x W) | 75 x 75 mm |
---|---|
Material | Polypropylene |
Packaging | Individually Bagged |
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
17584686
![]() |
Sampling Systems™
8785B-75 |
Non-sterile |
1345.00 DKK
Pack of 100 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
17594686
![]() |
Sampling Systems™
8785B-75S |
Sterile |
1595.00 DKK
Pack of 100 |
Estimated Shipment: 15-08-2025
Log in to see stock. |
||||||
Description
- This range of blue disposable Scrapers are available sterile and non-sterile.
- They are made from high quality polymers.
- These scrapers are ideal in the manufacture of creams, ointments and pastes.
- The bright blue color means they can easily seen if they are left in the product by mistake.
- SteriWare™ Scrapers are available sterile and non-sterile.
- Moulded and packed in a cleanroom.
- Individually bagged.
- Made form FDA and EU conforming materials.
- BSE/TSE free.
- Full Batch Traceability.
- Available as gamma irradiated sterile (SAL 10-6).
Specifications
75 x 75 mm | |
Polypropylene | |
Individually Bagged |