Learn More
SI Analytics™ BlueLine pH Combination Electrode with Conical Membrane
Ensure an accurate and fast measurement with precision manufacturing and high quality material. SI Analytics™ BlueLine pH Combination Electrode with Conical Membrane is for use with more critical measurements.
Brand:  SI Analytics™ 285129114
 View more versions of this product
Product Code. 12048083
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Available in a variety of connection types
  • Liquid electrolyte electrode
  • For demanding measurements
  • 120mm shaft length
  • Type A glass conical membrane
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plug Head
Glass
Combination pH Electrode
Cone
-5°C to +100°C
12 mm
Platinum Diaphragm
0 to 14
Ag/AgCl
Blueline 11 PH
120 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SI Analytics™ BlueLine pH Combination Electrode with Conical Membrane > Connector Type: Plug Head