missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS CCR2 22, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
High affinity CCR2 chemokine receptor antagonist (IC50 = 5.1 nM). Displays potent functional antagonism, IC50 values are 1 and 18 nM for chemotaxis and antagonism of calcium flux respectively. Selective over CCR3.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 593.66 |
Percent Purity | >98% |
Chemical Name or Material | BMS CCR2 22 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'