Learn More
Bohlender™ BOLA™ PTFE Tubing Connectors
Connect elastic tubing with these tubing connectors. BOLA PTFE Tubing Connectors feature universal chemical resistance and are vacuum suitable.
Brand:  Bohlender™ D575-06
 View more versions of this product
Product Code. 10737532
  • 161.00 DKK / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • PTFE straight fitting
  • Three connectors for elastic tubing (e.g. Viton, Tygon, silicone)
  • Universal chemical resistance
  • Product is only exposed to PTFE
Compliance
FDA
Specifications
Specifications
5 mm
6.8 mm
61 mm
6.8mm O.D., 5mm I.D.