Learn More
Bohlender™ BOLA™ Tri-clamp Gaskets
Double-sided tri-clamp gaskets with elastic PTFE sleeve and silicone inlay.
165.00 DKK - 391.00 DKK
Product Code | Brand | Diameter (Metric) Outside | For Use With (Equipment) | Size (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | For Use With (Equipment) | Size (Metric) | Price | Quantity & Availability | ||||
17187334
|
Bohlender™
D930-04 |
25 mm | Tubing with 10 mm I.D. | DN 10 |
165.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17197334
|
Bohlender™
D930-22 |
34 mm | Tubing with 10 mm I.D. | DN 10 |
186.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17107344
|
Bohlender™
D930-24 |
34 mm | Tubing with 16 mm I.D. | DN 15 |
181.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17117344
|
Bohlender™
D930-26 |
34 mm | Tubing with 20 mm I.D. | DN 20 |
179.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17127344
|
Bohlender™
D930-42 |
50.5 mm | Tubing with 26 mm I.D. | DN 25 |
211.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17137344
|
Bohlender™
D930-44 |
50.5 mm | Tubing with 32 mm I.D. | DN 32 |
211.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17147344
|
Bohlender™
D930-46 |
50.5 mm | Tubing with 38 mm I.D. | DN 40 |
207.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17157344
|
Bohlender™
D930-62 |
64 mm | Tubing with 50 mm I.D. | DN 50 |
367.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
17167344
|
Bohlender™
D930-72 |
91 mm | Tubing with 66 mm I.D. | DN 65 |
391.00 DKK
Each |
Estimated Shipment: 08-08-2025
Log in to see stock. |
||||
Description
- Silicone ring with double-sided, elastic washer made of PTFE
- Suitable for Tri-clamp connections as per DIN 32 676
- Universal chemical resistance
- Product is only exposed to PTFE
Specifications
PTFE, Silicone |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ BOLA™ Tri-clamp Gaskets