missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Unit Size:
1kg
2.50kg
25kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Vapor Pressure | 2.6mmHg (20°C) |
Quantity | 6 x 1 kg |
Linear Formula | H3BO3 |
UN Number | NONH for all modes of transport |
Solubility Information | In accordance with ethanol |
Formula Weight | 61.83 |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. |
Chemical Name or Material | Boric Acid, Meets analytical specification of Ph. Eur.BP, NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric acid, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.5-100.5%, powder, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction