missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
CAS | 10043-35-3 |
Quantity | 1 kg |
Molecular Formula | BH3O3 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
SMILES | OB(O)O |
IUPAC Name | boric acid |
Molecular Weight (g/mol) | 61.83 |
Grade | Reagent |
Safety and Handling

Product Identifier
- Boric acid, reagent grade, ACS, ISO, Reag. Ph Eur
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric acid, Scharlab >
Spot an opportunity for improvement?Share a Content Correction