Learn More
Boron Standard Metal Solution 10,000ppm, for ICP, Solution NH3 1%, Fisher Chemical
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
MDL Number | 134034 |
Physical Form | Liquid |