Learn More
Jaytec Glass™ Borosilicate Glass Butyrometer Tube
The Borosilicate Glass Butyrometer Tube is used to determine the fat content of dairy samples using the Gerber method.
Brand:  Jaytec Glass™ MP893
 View more versions of this product
Product Code. 12378955
  • 322.00 DKK / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
  • Borosilicate glass
  • Capacity and tolerances in accordance with BS ISO 488:2008 (formerly BS 696)
  • Graduated round stem
Specifications
Specifications
Borosilicate Glass
Milk Butyrometer
0.1%
10.94 mL
Determination of fat content using Gerber method
0 to 8%
Documents
One moment while we fetch your results.
Product Title
Jaytec Glass™ Borosilicate Glass Butyrometer Tube > Range: 0 to 8%