Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Gravity chromatography column manufactured from medium wall tubing with 821000 PTFE stopcock
Brand:  Kimble Chase™ 420540-0430
2990.67 DKK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 11857423
  • 3255.00 DKK / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Description
  • 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ion exchange chromatography
38 mm
Type I
300 mm
With fritted disc and PTFE stopcock plug
1 Each
Chromatography Column
340 mL
Borosilicate Glass
Product Title
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug > Column Capacity: 340mL