Learn More
IKA™ Borosilicate Glass RV 10.5 Reflux Distill Vertical Condensers
Brand:  IKA™ 0003743000
 View more versions of this product
Product Code. 11875932
  • 16473.80 DKK / Each
Only null left
Estimated Shipment: 07-10-2025
Specifications
Borosilicate Glass
Without Coating
1 Each
Reflux Distill Condenser Vertical
SURLYN
-30°C to +60°C
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ Borosilicate Glass RV 10.5 Reflux Distill Vertical Condensers > Without Coating