Learn More
IKA™ Borosilicate Glass RV 10.6 Reflux Distill Vertical Condensers
18085.37 DKK - 19711.86 DKK
Specifications
Material | Borosilicate Glass |
---|---|
Material (Coating) | SURLYN |
Quantity | 1 Each |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
11865942
|
IKA™
0003744100 |
Coating |
19711.86 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||||
11855942
|
IKA™
0003744000 |
Without Coating |
18085.37 DKK
Each |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||||||
Description
Specifications
Borosilicate Glass | |
SURLYN | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
IKA™ Borosilicate Glass RV 10.6 Reflux Distill Vertical Condensers