Learn More
Reitenspiess™ Bottle Brush
Clean, disinfect and sterilize your laboratory bottles using the Reitenspiess™ Bottle Brush. With a range of lengths and bristle types, the brush can be specified to your specific glassware needs.
Brand: Reitenspiess™ 22550234
42.28 DKK valid until 2025-05-18
Use promo code "24096" to get your promotional price.
Description
- Variety of bristle types
- 1 each in package
- Variety of lengths and diameters
Specifications
Bottle Brush | |
55 mm | |
1 Ea. |
Hog Bristle | |
500 mm |
Special Offers
-
Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.