Learn More
Reitenspiess™ Bottle Brush
Clean, disinfect and sterilize your laboratory bottles using the Reitenspiess™ Bottle Brush. With a range of lengths and bristle types, the brush can be specified to your specific glassware needs.
Brand:  Reitenspiess™ 63800234
 View more versions of this product
Product Code. 11301275
  • 223.00 DKK / Each
Only null left
Estimated Shipment: 20-08-2025
Description
  • Variety of bristle types
  • 1 each in package
  • Variety of lengths and diameters
Specifications
Specifications
Bottle Brush
80 mm
1 Ea.
Bleached Bristle
550 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Reitenspiess™ Bottle Brush > Length: 550mm