Learn More
Socorex™ Bottle Top Dispenser 530 Model
Brand:  Socorex™ 530.050
 View more versions of this product
Product Code. 13578290
  • 4550.00 DKK / Each
Only null left
Estimated Shipment: 09-10-2025
Specifications
Bottletop Dispenser
EN ISO 8655
530
Autoclavable
5 to 50 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Socorex™ Bottle Top Dispenser 530 Model > Volume Range: 5 to 50mL