Learn More
Trajan Scientific and Medical™ BP5 GC Capillary Columns
Used for a wide range of applications. Trajan Scientific and Medical BP5 GC Capillary Columns are general purpose phase columns.
Brand:  Trajan Scientific and Medical™ 054195
 View more versions of this product
Product Code. 15288653
  • 5945.00 DKK / Each
Estimated Shipment: 13-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Excellent general purpose column
  • Low bleed
  • Non-polar High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Specifications
0.53 mm
Hydrocarbons Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons, GC/MS
1 μm
5% Phenyl, 95% Dimethyl Polysiloxane
BP5 GC Capillary Column
-60°C to +320/340°C
30 m
BP5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ BP5 GC Capillary Columns > Length: 30m; Dia.: 0.53mm; Thickness: 1μm