Learn More
Brady™ B-7569 Pre-Printed GHS/CLP Chemical Labels, L4A Diamond
Brand: Brady™ 121637
This item is not returnable.
View return policy
Specifications
B30-260-7569-CLP4A | |
Chemical labeling | |
340/Roll |
Legend: L4A Diamond | |
BBP30, BBP31, BBP33, BBP35, and BBP37 printers | |
55 x 85 mm (2.17 x 3.35 in.) |