Learn More
Brady™ Indoor/Outdoor Vinyl Tape for B30 Printers
Durable, low-shrink Indoor/Outdoor Vinyl Tape with very aggressive adhesive.
949.00 DKK - 3405.00 DKK
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance RoHS 2002/95/EC
Description Indoor/Outdoor Vinyl Tape for Brady B30 Printers
Adhesive Type Permanent Acrylic, Permanent
Coating None
View More Specs
Products 38
Product Code Brand Color Width (English) Width (Metric) Price Quantity & Availability  
Product Code Brand Color Width (English) Width (Metric) Price Quantity & Availability  
11786635
View Documents
Brady™
142032
Yellow 4 in. 101.60 mm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11796635
View Documents
Brady™
142033
Yellow 3 in. 76.20 mm
3025.00 DKK
Each
In Stock
to see stock.
Add to basket
11716645
View Documents
Brady™
142035
Yellow 1.125 in. 28.58 mm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11706645
View Documents
Brady™
B30C-2250-595-YL
Yellow 2.25 in. 15.15 mm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11726645
View Documents
Brady™
142036
Yellow 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11776635
View Documents
Brady™
142031
White 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11746635
View Documents
Brady™
B30C-3000-595-WT
White 3 in. 7.62 cm
3025.00 DKK
Each
In Stock
to see stock.
Add to basket
11756635
View Documents
Brady™
B30C-2250-595-WT
White 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11766635
View Documents
Brady™
B30C-1125-595-WT
White 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11706635
View Documents
Brady™
B30C-2250-595-RD
Red 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11726635
View Documents
Brady™
142026
Red 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11736635
View Documents
Brady™
B30C-4000-595-WT
White 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11716635
View Documents
Brady™
142025
Red 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11796625
View Documents
Brady™
B30C-4000-595-RD
Red 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11746625
View Documents
Brady™
142018
Orange 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11726625
View Documents
Brady™
B30C-2250-595-OR
Orange 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11736625
View Documents
Brady™
142017
Orange 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11716625
View Documents
Brady™
142015
Orange 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11706625
View Documents
Brady™
142014
Gray 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11786615
View Documents
Brady™
B30C-2250-595-GY
Gray 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11796615
View Documents
Brady™
B30C-1125-595-GY
Gray 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11776615
View Documents
Brady™
142011
Gray 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11756615
View Documents
Brady™
142009
Green 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11746615
View Documents
Brady™
142008
Green 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11746495
View Documents
Brady™
142000
Clear 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11736615
View Documents
Brady™
142007
Green 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11766615
View Documents
Brady™
142010
Green 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11736495
View Documents
Brady™
141999
Clear 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11766495
View Documents
Brady™
142002
Clear 0.5 in. 12.70 mm
949.00 DKK
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
11756495
View Documents
Brady™
142001
Clear 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11786485
View Documents
Brady™
141994
Blue 0.5 in. 12.70 mm
949.00 DKK
Each
In Stock
to see stock.
Add to basket
11746485
View Documents
Brady™
141990
Black 0.5 in. 12.70 mm
949.00 DKK
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
11766485
View Documents
Brady™
141992
Blue 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11716485
View Documents
Brady™
141987
Black 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11756485
View Documents
Brady™
141991
Blue 4 in. 10.16 cm
3405.00 DKK
Each
In Stock
to see stock.
Add to basket
11726485
View Documents
Brady™
141988
Black 2.25 in. 5.72 cm
2275.00 DKK
Each
In Stock
to see stock.
Add to basket
11736485
View Documents
Brady™
141989
Black 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
11776485
View Documents
Brady™
141993
Blue 1.125 in. 2.86 cm
1515.00 DKK
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Conforms to irregular, curved, rough and highly textured surfaces.
  • Adheres to difficult low-surface energy items.
  • Can be applied at cold temperatures.
  • Length: 30.5 m (100 ft.) per roll.
Specifications
Specifications
Not Autoclavable
Indoor/Outdoor Vinyl Tape for Brady B30 Printers
None
Vinyl
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40°F to +179.6°F
0.004 in.
100 ft.
RoHS 2002/95/EC
Permanent Acrylic, Permanent
Gloss
Thermal Transfer
Pipe marking, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
50 Gardner Units
8 to 10 Years
Rectangle
-40°C to +82°C
0.102 mm
30.48 m