Learn More
Brady™ FreezerBondz II Polyester Laboratory Labels B490
FreezerBondz II Matte White Polyester Laboratory Labels are durable polyester markers designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 142893
 View more versions of this product
Product Code. 15313744
  • 938.00 DKK / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • White polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Brady Series R4300 ribbon is recommended
Specifications
Specifications
RoHS
Permanent Acrylic
Matte
Laboratory Label
Use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
1
LabelMark, MarkWare
-195 to 130°C
0.053 mm
0.3 in.
1.25 in.
FreezerBondz Labels for the BBP33 Printer
White
Polyester
Non-sterile
BBP™33 Printer
1500/Pk.
-320 to 266°F
0.0021 in.
2 x 1.25 in.
31.75 mm
3.17 cm